Merck
  • Simultaneous discrimination of diameter, handedness, and metallicity of single-walled carbon nanotubes with chiral diporphyrin nanocalipers.
Simultaneous discrimination of diameter, handedness, and metallicity of single-walled carbon nanotubes with chiral diporphyrin nanocalipers.
Journal of the American Chemical Society (2013-03-05)
Gang Liu, Feng Wang, Songpol Chaunchaiyakul, Yukie Saito, Ajoy K Bauri, Takahide Kimura, Yuji Kuwahara, Naoki Komatsu
ABSTRACT
We have been developing the methodology to discriminate the handedness and diameter of single-walled carbon nanotubes (SWNTs) through molecular recognition using chiral diporphyrin nanotweezers. Although relatively small diameters of SWNTs (<1.0 nm) were recognized well, nanotweezers were not able to form stable complexes with the SWNTs having the diameters >1.0 nm. In this context, we designed chiral diporphyrin with a much larger cavity, namely, "nanocalipers". The feature of the newly designed host molecule is: (1) long spacer with more than 1.4 nm consisting of three aromatic moieties; (2) nearly parallel orientation of the two porphyrins; (3) restricted conformation by biaryl linkages of the porphyrin-carbazole and carbazole-anthracene; (4) strong interaction of two porphyrins and anthracene with the surface of a SWNT through π-π stacking; and (5) stereogenic centers at the periphery of porphyrins discriminating helicity of SWNTs. As expected, we obtained optically active SWNTs with >1.0 nm in diameter and, unexpectedly, enriched metallic SWNTs over semiconducting ones. The optically active metallic SWNTs are identified for the first time, in addition to the optically active semiconducting SWNTs with such large diameters. The nanocalipers are found to recognize the diameter, handedness, and metallicity of SWNTs simultaneously.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Carbazole, VETRANAL®, analytical standard
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Carbazole, ≥95% (GC)
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, reagent grade, 97%