Merck
  • High-density lipoprotein subfractions--what the clinicians need to know.
High-density lipoprotein subfractions--what the clinicians need to know.
Cardiology (2013-02-23)
Angela Pirillo, Giuseppe Danilo Norata, Alberico Luigi Catapano
ABSTRACT
Although the inverse relationship between plasma levels of high-density lipoprotein (HDL) and cardiovascular disease has been largely demonstrated, many observations have suggested that the assessment of HDL functionality might be more informative than a simple measurement of HDL-cholesterol plasma levels. HDLs are a class of structurally and functionally heterogeneous particles; in atherosclerosis-related diseases, changes in HDL subfraction levels and functions are frequently observed. Circulating levels of large HDL particles are decreased in dyslipidaemic conditions, while levels of small dense HDL particles are increased in patients with coronary heart disease. Furthermore, specific genetic defects in proteins involved in HDL metabolism significantly impact the distribution of HDL subpopulations. Finally, many drugs used for dyslipidaemia induce changes in HDL subfractions strictly related to cardiovascular disease. Although several methods exist to evaluate HDL subclass levels, most of them are not easily applicable in clinical practice, due to the costs and high variability. However, the possibility to measure the levels of specific HDL subfractions in patients with atherosclerosis-related diseases might help to better define their cardiovascular risk.
Product Description
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid sodium salt, 98%
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material