Merck
  • Case report: analysis of an ultra-high-molecular-weight polyethylene acetabular cup retrieval at 41 years.
Case report: analysis of an ultra-high-molecular-weight polyethylene acetabular cup retrieval at 41 years.
Journal of long-term effects of medical implants (2013-02-23)
Frederick J Kummer, Lizeth Herrera, Alec Weir, Kim-Phuong Le, William L Jaffe
ABSTRACT
A polyethylene cup explanted after 41 years was examined using several analytical techniques to determine whether there was a material cause for the extremely low wear observed. Neither the amount of polyethylene oxidation nor crystallinity appeared to be a factor.
Product Description
Polyethylene (LDPE), ERM®, certified reference material
Supelco
Polyethylene, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)