- [Nasal drops addiction--the case report].
[Nasal drops addiction--the case report].
Przeglad lekarski (2013-02-21)
Katarzyna Korzeniowska, Karolina Simon, Anna Jabłecka
PMID23421118
ABSTRACT
The article describes the case of 34-years old man, who has used nasal drops with xylomethazoline for three years. Health consequence of uncontrolled use of the drops and treatment were prescribed. Described problem confirms the need of physicians and pharmacists cooperation to limit the problem of drug-addiction.
MATERIALS