Merck
  • Antioxidant synergism between ethanolic Centella asiatica extracts and α-tocopherol in model systems.
Antioxidant synergism between ethanolic Centella asiatica extracts and α-tocopherol in model systems.
Food chemistry (2013-02-16)
Yin Yin Thoo, Faridah Abas, Oi-Ming Lai, Chun Wai Ho, Jie Yin, Rikke V Hedegaard, Leif H Skibsted, Chin Ping Tan
ABSTRACT
The synergistic antioxidant effects of ethanolic extracts of Centella asiatica (CE), and α-tocopherol have been studied. The types of interactions exhibited by CE and α-tocopherol combined at different ratios were measured using three assays: 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) diammonium salt (ABTS) radical-scavenging capacity, the β-carotene bleaching system and liposome peroxidation assays. Fixed-fraction isobolographic analysis was used to detect any inducement of the antioxidant activity compared with the individual activities of CE and α-tocopherol. Of all synergistic combinations of CE and α-tocopherol, only fraction 2/3 showed the synergistic combination that fits well in three different assays and can be explained by the regeneration of α-tocopherol by CE despite the interaction effect of β-carotene present in the analytical assay. This phenomenon involved complex interactions between CE and α-tocopherol to exhibit different degrees of interactions that eventually increased antioxidant activity.
Product Description
Supelco
DL-alpha-Tocopherol acetate, analytical standard
Sigma-Aldrich
DL-α-Tocopherol acetate, tested according to Ph. Eur.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
α-Tocopherol acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Tocopheryl Acetate, a, Pharmaceutical Secondary Standard; Certified Reference Material
All-rac-alpha-tocopheryl acetate for peak identification, European Pharmacopoeia (EP) Reference Standard