Merck
  • Deformation properties of pharmaceutical excipients determined using an in-die and out-die method.
Deformation properties of pharmaceutical excipients determined using an in-die and out-die method.
International journal of pharmaceutics (2013-02-14)
Ilija Ilić, Biljana Govedarica, Rok Šibanc, Rok Dreu, Stane Srčič
ABSTRACT
This study investigated deformation mechanisms of some commonly used pharmaceutical fillers, such as microcrystalline cellulose, lactose, dicalcium phosphate, isomalt and cornstarch, using a combination of the in-die and out-die method with the Heckel and Walker models. The tableting mixtures contained of 98.5% (w/w) filler, the rest consisted of dry binder and an antiadhesive agent. Our results showed that plasticity and elasticity may be considered independent deformation properties as highly plastic materials (microcrystalline cellulose, cornstarch) also exhibited high elasticity. Particular emphasis was placed on explaining the differences observed between the in-die and out-die method-comparison revealed that the differences are a consequence of the material's elastic properties. Larger error of in-die results can be expected for more elastic materials, and thus in-die Heckel should be used with some considerations. In contrast, the Walker model was found to be more robust and smaller differences were observed between the two methods. We consider the most correct results to have been obtained by the out-die approach, which excludes the elastic properties of the material evaluated. An excellent correlation between elastic determination at the single-particle level and multiple-particle scale was demonstrated, suggesting a great potential of nanoscale determination of a material's mechanical properties for better elucidation of deformation mechanisms.
Product Description
Isomalt, European Pharmacopoeia (EP) Reference Standard
USP
Isomalt, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material