Merck
  • Acetosolv pulping for the fractionation of empty fruit bunches from palm oil industry.
Acetosolv pulping for the fractionation of empty fruit bunches from palm oil industry.
Bioresource technology (2013-02-12)
A Ferrer, A Vega, A Rodríguez, L Jiménez
ABSTRACT
Influence of operational variables in the EFB pulping [acetic acid (60-95%), hydrochloric acid (0.10-0.25%) and time (60-180 min)], on the yield, drainability or beating grade, Kappa number, lignin and viscosity of pulps was studied. By using a factorial design, equations were obtained that reproduced the experimental results for the dependent variables with errors less than 9-18%. These equations can be used to find the suitable conditions, so that operating with not too high values of operating variables (with minor costs of operation and capital) pulps with acceptable properties could be obtained: operating with 86.25% acetic acid, 0.25% hydrochloric acid and 120 min time, produced pulps with 46.56% yield, 15.9 °SR drainability, 36.3 Kappa number, 10.3% lignin and 303 mL/g viscosity, values all of them close to the optimal predicted.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Palm oil, analytical standard
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid