Merck
  • Synthesis and biological evaluation of 1-(2-hydroxy-3-phenyloxypropyl)piperazine derivatives as T-type calcium channel blockers.
Synthesis and biological evaluation of 1-(2-hydroxy-3-phenyloxypropyl)piperazine derivatives as T-type calcium channel blockers.
Bioorganic & medicinal chemistry letters (2013-02-12)
Jung-Eun Park, Wan Keun Ji, Jae Wan Jang, Ae Nim Pae, Keehyun Choi, Ki Hang Choi, Jahyo Kang, Eun Joo Roh
ABSTRACT
To obtain selective and potent inhibitor for T-type calcium channel by ligand based drug design, 2-hydroxy-3-phenoxypropyl piperazine derivatives were synthesized and evaluated for in vitro activities. Compound 6m and 6q showed high selectivity over hERG channel (IC50 ratio of hERG/α1G6m=8.5, 6q=18.38) and they were subjected to measure pharmacokinetics profiles. Among them compound 6m showed an excellent pharmacokinetic profile in rats.
Product Description
Supelco
Piperazine hexahydrate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Pricing and availability is not currently available.