Merck
Measurement of the thermal relaxation time in agar-gelled water.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2013-02-01)
Raphael H Matsunaga, Icaro dos Santos
ABSTRACT
In this study, we presented an experiment to obtain the thermal relaxation time which is necessary to model heat conduction by the hyperbolic heat equation. This experiment was evaluated by finite element simulation to acquire reliably this parameter for biological tissue. Besides that, we measured the thermal relaxation time of agar-gelled water with 2% of concentration at 25°C. The average value of thermal relaxation time for the gel was 7.9630s with standard deviation of 1.4562.
Product Description
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Select agar
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Bacteriological agar, for molecular biology