Merck
  • Pt-like behavior of high-performance counter electrodes prepared from binary tantalum compounds showing high electrocatalytic activity for dye-sensitized solar cells.
Pt-like behavior of high-performance counter electrodes prepared from binary tantalum compounds showing high electrocatalytic activity for dye-sensitized solar cells.
ChemSusChem (2013-01-31)
Sining Yun, Mingxing Wu, Yudi Wang, Jing Shi, Xiao Lin, Anders Hagfeldt, Tingli Ma
ABSTRACT
Ta-based compounds show Pt-like behavior: Binary tantalum compounds as counter electrodes (CEs) in dye-sensitized solar cells (DSCs) demonstrate Pt-like electrocatalytic activity and competitive photovoltaic performance, matching the performance of DSCs with Pt CEs. The first-principle density functional theory (DFT) calculations provide a strategy for understanding the relationship between the electronic structure and the catalytic activity of CE catalysts in DSCs.
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis