Merck
  • Effect of in vivo chronic exposure to clotrimazole on zebrafish testis function.
Effect of in vivo chronic exposure to clotrimazole on zebrafish testis function.
Environmental science and pollution research international (2013-01-24)
Damien Baudiffier, Nathalie Hinfray, Catherine Ravaud, Nicolas Creusot, Edith Chadili, Jean-Marc Porcher, Rüdiger W Schulz, François Brion
ABSTRACT
Clotrimazole is an azole fungicide used as a human pharmaceutical that is known to inhibit cytochrome P450 (CYP) enzymatic activities, including several steroidogenic CYP. In a previous report, we showed that a 7-day exposure to clotrimazole induced the expression of genes related to steroidogenesis in the testes as a compensatory response, involving the activation of the Fsh/Fshr pathway. In this context, the aim of the present study was to assess the effect of an in vivo 21-day chronic exposure to clotrimazole (30-197 μg/L) on zebrafish testis function, i.e., spermatogenesis and androgen release. The experimental design combined (1) gene transcript levels measurements along the brain-pituitary-gonad axis, (2) 11-ketotestosterone (11-KT) quantification in the blood, and (3) histology of the testes, including morphometric analysis. The chronic exposure led to an induction of steroidogenesis-related genes and fshr in the testes as well as fshβ in the pituitary. Moreover, increases of the gonadosomatic index and of the volume proportion of interstitial Leydig cells were observed in clotrimazole-exposed fish. In accordance with these histological observations, the circulating concentration of 11-KT had increased. Morphometric analysis of the testes did not show an effect of clotrimazole on meiotic (spermatocytes) or postmeiotic (spermatids and spermatozoa) stages, but we observed an increase in the number of type A spermatogonia, in agreement with an increase in mRNA levels of piwil1, a specific molecular marker of type A spermatogonia. Our study demonstrated that clotrimazole is able to affect testicular physiology and raised further concern about the impact of clotrimazole on reproduction.
Sigma-Aldrich
11-Ketotestosterone
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Clotrimazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clotrimazole
Clotrimazole, European Pharmacopoeia (EP) Reference Standard