Merck
  • Genetic polymorphisms in catalase and CYP1B1 determine DNA adduct formation by benzo(a)pyrene ex vivo.
Genetic polymorphisms in catalase and CYP1B1 determine DNA adduct formation by benzo(a)pyrene ex vivo.
Mutagenesis (2013-01-18)
Marten A Schults, Roland K Chiu, Peter W Nagle, Lonneke C Wilms, Jos C Kleinjans, Frederik J van Schooten, Roger W Godschalk
ABSTRACT
Genetic polymorphisms can partially explain the large inter-individual variation in DNA adduct levels following exposure to polycyclic aromatic hydrocarbons. Effects of genetic polymorphisms on DNA adduct formation are difficult to assess in human studies because exposure misclassification attenuates underlying relationships. Conversely, ex vivo studies offer the advantage of controlled exposure settings, allowing the possibility to better elucidate genotype-phenotype relationships and gene-gene interactions. Therefore, we exposed lymphocytes of 168 non-smoking volunteers ex vivo to the environmental pollutant benzo(a)pyrene (BaP) and BaP-related DNA adducts were quantified. Thirty-four genetic polymorphisms were assessed in genes involved in carcinogen metabolism, oxidative stress and DNA repair. Polymorphisms in catalase (CAT, rs1001179) and cytochrome P450 1B1 (CYP1B1, rs1800440) were significantly associated with DNA adduct levels, especially when combined. Moreover, reverse transcription-polymerase chain reaction (RT-PCR) analysis in a subset of 30 subjects revealed that expression of catalase correlated strongly with expression of CYP1B1 (R = 0.92, P < 0.001). To further investigate the mechanism by which catalase influences CYP1B1 and how they simultaneously affect BaP-related DNA adduct levels, catalase expression was transiently knocked down in the human lung epithelial cell line A549. Although catalase knockdown did not immediately change CYP1B1 gene expression, recovery of catalase expression 8 h after the knockdown coincided with a 2.2-fold increased expression of CYP1B1 (P < 0.05). We conclude that the genetic polymorphism in the promoter region of CAT may determine the amount and activity of catalase, which may subsequently regulate the expression of CYP1B1. As a result, both genetic polymorphisms modulate DNA adduct levels in lymphocytes by BaP ex vivo.
Product Description
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland