Merck
  • Cu(OTf)2-catalyzed synthesis of 2,3-disubstituted indoles and 2,4,5-trisubstituted pyrroles from α-diazoketones.
Cu(OTf)2-catalyzed synthesis of 2,3-disubstituted indoles and 2,4,5-trisubstituted pyrroles from α-diazoketones.
Organic letters (2013-01-18)
B V Subba Reddy, M Ramana Reddy, Y Gopal Rao, J S Yadav, B Sridhar
ABSTRACT
A novel method has been devised for the synthesis of 2,4,5-trisubstituted pyrrole derivatives through the coupling of α-diazoketones with β-enaminoketones and esters using 10 mol % of Cu(OTf)(2). A wide range of 2,3-disubstituted indole derivatives were also prepared from α-diazoketones and 2-aminoaryl or alkyl ketones. The synthetic versatility of this approach has been exemplified in the formal synthesis of homofascaplysin C.
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis