Merck
  • NIR- and FRET-based sensing of Cu2+ and S2- in physiological conditions and in live cells.
NIR- and FRET-based sensing of Cu2+ and S2- in physiological conditions and in live cells.
Inorganic chemistry (2013-01-11)
Chirantan Kar, Manab Deb Adhikari, Aiyagari Ramesh, Gopal Das
ABSTRACT
We have synthesized a new indole functionalized rhodamine derivative L(1) which specifically binds to Cu(2+) in the presence of large excess of other competing ions with visually observable changes in their electronic and fluorescence spectral behavior. These spectral changes are significant enough in the NIR and visible region of the spectrum and thus enable naked eye detection. The receptor, L(1), could be employed as a resonance energy transfer (RET) based sensor for detection of Cu(2+) based on the process involving the donor indole and the acceptor Cu(2+) bound xanthene fragment. Studies reveal that L(1)-Cu complex is selectively and fully reversible in presence of sulfide anions. Further, fluorescence microscopic studies confirmed that the reagent L(1) could also be used as an imaging probe for detection of uptake of these ions in HeLa cells.
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol