Merck
  • Significant increases in potato Starch-Synthase and Starch-Branching-Enzyme activities by dilution with buffer containing dithiothreitol and polyvinyl alcohol 50 K.
Significant increases in potato Starch-Synthase and Starch-Branching-Enzyme activities by dilution with buffer containing dithiothreitol and polyvinyl alcohol 50 K.
Carbohydrate research (2013-01-09)
Rupendra Mukerjea, Alexander N Gray, John F Robyt
ABSTRACT
We have recently found that the dilution of purified potato Starch-Synthases (SS) and Starch-Branching-Enzymes (SBEs), by a glycine buffer (pH 8.5), containing 1.0mM dithiothreitol (DTT) and 0.04% (w/v) polyvinyl alcohol (PVA) 50K, produced a striking and significant increase in activity (mIU/mL and total mIU) when diluted 1→2 to 1→10. As an example, one SS fraction diluted 1→10 went from 259 to 1470 mIU/mL, giving a total of 14,700 mIU. Dilutions of 1→15 usually resulted in a complete loss of activity. Removal of both DTT and PVA also gave the complete loss of activity. Individual removal of just DTT and the removal of just PVA also produced lowered activities on dilution. The addition of the DTT and the PVA back to the diluted fractions did produce an increase in the activity, but never to the extent that occurred when the samples were diluted simultaneously with both DTT and PVA together in the diluting buffer. Dilution of SBE with buffer containing both DTT and PVA, gave moderate increases, with the exception of one fraction that diluted 1→20 gave a significant increase from 18 to 382 mIU/mL and a total of 7640 mIU. It is concluded that there are inactive starch synthesizing enzymes in the purified fractions that are significantly activated by DTT and PVA, giving much greater amounts of enzyme activities.
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000