Merck
  • Evaluation of three selective chromogenic media, CHROMagar ESBL, CHROMagar CTX-M and CHROMagar KPC, for the detection of Klebsiella pneumoniae producing OXA-48 carbapenemase.
Evaluation of three selective chromogenic media, CHROMagar ESBL, CHROMagar CTX-M and CHROMagar KPC, for the detection of Klebsiella pneumoniae producing OXA-48 carbapenemase.
Journal of clinical pathology (2013-01-05)
Michael Hornsey, Lynette Phee, Neil Woodford, Jane Turton, Daniele Meunier, Claire Thomas, David W Wareham
ABSTRACT
Three selective chromogenic culture media (CHROMagars ESBL, CTX-M and KPC) were evaluated for their ability to support the growth of nine Klebsiella pneumoniae isolates producing OXA-48 carbapenemase in combination with other β-lactamases. CHROMagar ESBL and CHROMagar KPC were the most sensitive media, supporting growth of all isolates with a detection limit as low as < 100 CFU/ml. Five isolates failed to grow on CHROMagar CTX-M, and five were recovered on CHROMagar KPC only at counts > 10(6) CFU/ml. Both CHROMagar ESBL and CHROMagar KPC may be useful for enhanced isolation of K pneumoniae producing OXA-48-like carbapenemases.
Product Description
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, suitable for plant cell culture
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Bacteriological agar, for molecular biology