Merck
  • Synthesis and in vitro activity of 1,2,4-triazole-ciprofloxacin hybrids against drug-susceptible and drug-resistant bacteria.
Synthesis and in vitro activity of 1,2,4-triazole-ciprofloxacin hybrids against drug-susceptible and drug-resistant bacteria.
European journal of medicinal chemistry (2013-01-05)
Tomasz Plech, Monika Wujec, Urszula Kosikowska, Anna Malm, Barbara Rajtar, Małgorzata Polz-Dacewicz
ABSTRACT
A series of novel 1,2,4-triazole-ciprofloxacin hybrids was designed, synthesised and evaluated in vitro against drug-susceptible and drug-resistant bacteria. A significant part of the compounds obtained showed antibacterial activity higher than the activity of ciprofloxacin, both towards Gram-positive and Gram-negative species. Despite relatively small number of synthesised derivatives, it was possible to observe important dependences between their structure and activity.
Sigma-Aldrich
1,2,4-Triazole, 98%
Pricing and availability is not currently available.