Merck
Placement of molecules in (not out of) the cell.
Acta crystallographica. Section D, Biological crystallography (2013-01-01)
Zbigniew Dauter
ABSTRACT
To uniquely describe a crystal structure, it is sufficient to specify the crystal unit cell and symmetry, and describe the unique structural motif which is repeated by the space-group symmetry throughout the whole crystal. It is somewhat arbitrary how such a unique motif can be defined and positioned with respect to the unit-cell origin. As a result of such freedom, some isomorphous structures are presented in the Protein Data Bank in different locations and appear as if they have different atomic coordinates, despite being completely equivalent structurally. This may easily confuse those users of the PDB who are less familiar with crystallographic symmetry transformations. It would therefore be beneficial for the community of PDB users to introduce standard rules for locating crystal structures of macromolecules in the unit cells of various space groups.
Sigma-Aldrich
Aprotinin from bovine lung
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg