Merck
  • Rac1 is a novel regulator of contraction-stimulated glucose uptake in skeletal muscle.
Rac1 is a novel regulator of contraction-stimulated glucose uptake in skeletal muscle.
Diabetes (2013-01-01)
Lykke Sylow, Thomas E Jensen, Maximilian Kleinert, Joshua R Mouatt, Stine J Maarbjerg, Jacob Jeppesen, Clara Prats, Tim T Chiu, Shlomit Boguslavsky, Amira Klip, Peter Schjerling, Erik A Richter
ABSTRACT
In skeletal muscle, the actin cytoskeleton-regulating GTPase, Rac1, is necessary for insulin-dependent GLUT4 translocation. Muscle contraction increases glucose transport and represents an alternative signaling pathway to insulin. Whether Rac1 is activated by muscle contraction and regulates contraction-induced glucose uptake is unknown. Therefore, we studied the effects of in vivo exercise and ex vivo muscle contractions on Rac1 signaling and its regulatory role in glucose uptake in mice and humans. Muscle Rac1-GTP binding was increased after exercise in mice (~60-100%) and humans (~40%), and this activation was AMP-activated protein kinase independent. Rac1 inhibition reduced contraction-stimulated glucose uptake in mouse muscle by 55% in soleus and by 20-58% in extensor digitorum longus (EDL; P < 0.01). In agreement, the contraction-stimulated increment in glucose uptake was decreased by 27% (P = 0.1) and 40% (P < 0.05) in soleus and EDL muscles, respectively, of muscle-specific inducible Rac1 knockout mice. Furthermore, depolymerization of the actin cytoskeleton decreased contraction-stimulated glucose uptake by 100% and 62% (P < 0.01) in soleus and EDL muscles, respectively. These are the first data to show that Rac1 is activated during muscle contraction in murine and human skeletal muscle and suggest that Rac1 and possibly the actin cytoskeleton are novel regulators of contraction-stimulated glucose uptake.
Sigma-Aldrich
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate, ≥93%
Supelco
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate, analytical standard
Sigma-Aldrich
Rac1 Inhibitor, Rac1 Inhibitor, CAS 1177865-17-6, is a cell-permeable, reversible inhibitor of Rac1 GDP/GTP exchange. Interferes with the interaction between Rac1 and Rac-specific GEFs Trio and Tiam1 (IC₅₀ ~50 µM).
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Rac1 Inhibitor II, The Rac1 Inhibitor II, also referenced under CAS 1090893-12-1, controls the biological activity of Rac1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid