Merck
  • Effect of acidic condition on the metabolic regulation of Escherichia coli and its phoB mutant.
Effect of acidic condition on the metabolic regulation of Escherichia coli and its phoB mutant.
Archives of microbiology (2013-01-01)
Lolo Wal Marzan, Chowdhury Mohammad Monirul Hasan, Kazuyuki Shimizu
ABSTRACT
Effect of acidic condition on the fermentation characteristics was investigated by the continuous culture of Escherichia coli. In accordance with down-regulation of crp gene transcript level as well as up-regulation of arcA, the expressions of the TCA cycle genes were down-regulated, which caused more acetate formation at acidic condition under aerobic condition. It was also shown that yfiD was up-regulated in accordance with up-regulation of fnr, and the respiratory pathway genes were up-regulated under acidic condition. The effect of phoB gene knockout on fermentation characteristics was also investigated. Under micro-aerobic condition, the fermentation pattern changed in such a way that formate and lactate were more produced at lower pH due to up-regulations of pflA, yfiD and ldhA genes, whereas ethanol was less produced as compared to the case at neutral pH. The overall regulation mechanism under acidic condition was clarified based on fermentation characteristics and gene transcript levels.
Product Description
Supelco
Calcium formate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%