- Effects of genotype and dietary oil supplementation on performance, carcass traits, pork quality and fatty acid composition of backfat and intramuscular fat.
Effects of genotype and dietary oil supplementation on performance, carcass traits, pork quality and fatty acid composition of backfat and intramuscular fat.
A 42-day study was conducted to evaluate the effect of genotype: terminal sire line Duroc×F1 (DC×F1); terminal sire line Embrapa MS-115×F1 (MS-115×F1); and MS-115×Moura (MS-115×MO) and three dietary oil sources: soybean; canola; and canola+flax, on performance, carcass traits, pork quality, and fatty acid composition. Genotype affected the technological quality of pork and fatty acid profile. MS-115-sired pigs had better meat color and Duroc-sired pigs had higher intramuscular fat content, more saturated fat and better omega-6/omega-3 ratio. Moura breed influenced positively meat tenderness and intramuscular fat. Diet did not affect the technological quality of the meat. Canola or canola+flax oil diet supplementations increased monounsaturated and C18:3 and decreased C18:2 fatty acids, reducing the omega-6/omega-3 ratio. The best omega-6/omega-3 ratio was obtained through supplementation with canola+flax.