Merck
Aflatoxin biosynthesis: current frontiers.
Annual review of food science and technology (2012-12-19)
Ludmila V Roze, Sung-Yong Hong, John E Linz
ABSTRACT
Aflatoxins are among the principal mycotoxins that contaminate economically important food and feed crops. Aflatoxin B1 is the most potent naturally occurring carcinogen known and is also an immunosuppressant. Occurrence of aflatoxins in crops has vast economic and human health impacts worldwide. Thus, the study of aflatoxin biosynthesis has become a focal point in attempts to reduce human exposure to aflatoxins. This review highlights recent advances in the field of aflatoxin biosynthesis and explores the functional connection between aflatoxin biosynthesis, endomembrane trafficking, and response to oxidative stress. Dissection of the regulatory mechanisms involves a complete comprehension of the aflatoxin biosynthetic process and the dynamic network of transcription factors that orchestrates coordinated expression of the target genes. Despite advancements in the field, development of a safe and effective multifaceted approach to solve the aflatoxin food contamination problem is still required.
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard
Supelco
Aflatoxin B1 solution, 2 μg/mL in acetonitrile, analytical standard