MilliporeSigma
  • RGD peptide-conjugated multimodal NaGdF4:Yb3+/Er3+ nanophosphors for upconversion luminescence, MR, and PET imaging of tumor angiogenesis.
RGD peptide-conjugated multimodal NaGdF4:Yb3+/Er3+ nanophosphors for upconversion luminescence, MR, and PET imaging of tumor angiogenesis.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2012-12-13)
Junghan Lee, Tae Sup Lee, Jiyoung Ryu, Sukmin Hong, Moonsik Kang, Kangbin Im, Joo Hyun Kang, Sang Moo Lim, Sun Park, Rita Song
ABSTRACT
Multimodal nanoparticles have been extensively studied for target-specific imaging and therapy of various diseases, including cancer. In this study, radiolabeled arginine-glycine-aspartic acid (RGD)-functionalized Er(3+)/Yb(3+) co-doped NaGdF(4) upconversion nanophosphors (UCNPs) were synthesized and evaluated as a multimodal PET/MR/optical probe with tumor angiogenesis-specific targeting properties. A dimeric cyclic RGDyk ((cRGDyk)(2)) peptide was conjugated to polyacrylic acid-coated NaGdF(4):Yb(3+)/Er(3+) UCNPs along with polyethylene glycol molecules and was consecutively radiolabeled with (124)I. In vitro cytotoxicity testing was performed for 3 d. Upconversion luminescence imaging of (cRGDyk)(2)-UCNP was performed on U87MG cells with a laboratory-made confocal microscope. In vivo small-animal PET and clinical 3-T T1-weighted MR imaging of (124)I-labeled RGD-functionalized UCNPs was acquired with or without blocking of cyclic RGD peptide in a U87MG tumor model. Inductively coupled plasma mass spectrometry and biologic transmission electron microscopy were done to evaluate gadolinium concentration and UCNP localization, respectively. Polymer-coated UCNPs and dimeric RGD-conjugated UCNPs were monodispersely synthesized, and those of hydrodynamic size were 30 ± 8 nm and 32 ± 9 nm, respectively. (cRGDyk)(2)-UCNPs have a low cytotoxic effect on cells. Upconversion luminescence signals of (cRGDyk)(2)-UCNP were specifically localized on the surface of U87MG cells. (124)I-c(RGDyk)(2)-UCNPs specifically accumulated in U87MG tumors (2.8 ± 0.8 vs. 1.3 ± 0.4 percentage injected dose per gram in the blocking experiment), and T1-weighted MR images showed significant positive contrast enhancement in U87MG tumors. Tumor localization of (124)I-c(RGDyk)(2)-UCNPs was confirmed by inductively coupled plasma mass spectrometry and biologic transmission electron microscopy analysis. These results suggest that (124)I-labeled RGD-functionalized UCNPs have high specificity for α(v)β(3) integrin-expressing U87MG tumor cells and xenografted tumor models. Multimodal UCNPs can be used as a platform nanoparticle with multimodal imaging for cancer-specific diagnoses.
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis