Merck
  • A nanosilver-based spectrophotometric method for determination of malachite green in surface water samples.
A nanosilver-based spectrophotometric method for determination of malachite green in surface water samples.
Environmental monitoring and assessment (2012-12-04)
R Sahraei, A Farmany, S S Mortazavi, H Noorizadeh
ABSTRACT
A new spectrophotometric method is reported for the determination of nanomolar level of malachite green in surface water samples. The method is based on the catalytic effect of silver nanoparticles on the oxidation of malachite green by hexacyanoferrate (III) in acetate-acetic acid medium. The absorbance is measured at 610 nm with the fixed-time method. Under the optimum conditions, the linear range was 8.0 × 10(-9)-2.0 × 10(-7) mol L(-1) malachite green with a correlation coefficient of 0.996. The limit of detection (S/N = 3) was 2.0 × 10(-9) mol L(-1). Relative standard deviation for ten replicate determinations of 1.0 × 10(-8) mol L(-1) malachite green was 1.86%. The method is featured with good accuracy and reproducibility for malachite green determination in surface water samples without any pre-concentration and separation step.
Sigma-Aldrich
Malachite Green solution, for microscopy
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, certified by the BSC
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade