Merck
  • Net toroidal magnetic moment in the ground state of a {Dy6}-triethanolamine ring.
Net toroidal magnetic moment in the ground state of a {Dy6}-triethanolamine ring.
Journal of the American Chemical Society (2012-11-01)
Liviu Ungur, Stuart K Langley, Thomas N Hooper, Boujemaa Moubaraki, Euan K Brechin, Keith S Murray, Liviu F Chibotaru
ABSTRACT
A toroidal magnetic moment in the absence of conventional total magnetic moment is observed in a {Dy(6)} ring. The reason for the net toroidal arrangement of the local magnetic moments is the high symmetry of the complex in combination with strong intra-molecular dipolar interactions between Dy ions. The description of single-ion and inter-ion anisotropic magnetic interactions is achieved here for the first time fully ab initio, i.e., without use of phenomenological parameters.
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine hydrochloride, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine hydrochloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)