Merck
  • Combined biophysical and cell-based approaches for the assessment of ligand binding to PPARγ.
Combined biophysical and cell-based approaches for the assessment of ligand binding to PPARγ.
Methods in molecular biology (Clifton, N.J.) (2012-10-27)
Silvia Zorrilla, Dolores Pérez-Sala
ABSTRACT
Transcription factors of the peroxisome proliferator-activated receptor (PPAR) family are ligand-activated receptors that play key roles in lipid metabolism and inflammation. The γ isoform (PPARγ) is involved in adipocyte differentiation, insulin sensitization, and vascular pathophysiology, including inflammation and atherosclerosis, for which it is considered an important drug target. PPARγ ligands display varied structures and include fatty acids, electrophilic lipids, and certain drugs. These agonists promote conformational changes allowing interaction of PPARγ with coactivators and hence transcriptional regulation. Here we present a panoply of methods to study PPARγ interactions with ligands and activation in vitro and in cells. The first method is based on the competition of the fluorescent dye 1-anilinonaphthalene-8-sulfonic acid (ANS) with PPARγ ligands for the ligand binding pocket, allowing detection and quantification of ligand binding to PPARγ. This method is specific for PPARγ while ANS displays negligible interaction with other nuclear receptors such as PPARα and retinoid X receptor α (RXRα). The ANS competition assay has been validated through comparison of the affinities determined for well-known PPARγ ligands by this method with those reported in the literature. We also describe here gel-based competition assays that show limited performance with non-covalently bound ligands. In addition, we present a fluorescence anisotropy assay to analyze PPARγ activation by ligands in vitro through their capacity of eliciting PPARγ interaction with a fluorescently labeled peptide derived from one of its coactivators (SRC-1). Finally, we show cell-based assays to investigate PPARγ activation by interaction with its ligands. We believe that combined approaches using ANS, fluorescent coactivator peptides, and in-cell assays to monitor PPARγ binding and interactions may provide valuable strategies for the identification and characterization of PPARγ ligands.
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95% (TLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95.0% (T)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, technical, ≥90% (NT)