Merck
  • Synthesis of indenes via Brønsted acid catalyzed cyclization of diaryl- and alkyl aryl-1,3-dienes.
Synthesis of indenes via Brønsted acid catalyzed cyclization of diaryl- and alkyl aryl-1,3-dienes.
Organic letters (2012-10-26)
Dahan Eom, Sangjune Park, Youngchul Park, Taekyu Ryu, Phil Ho Lee
ABSTRACT
Substituted indenes can be synthesized via the Brønsted acid catalyzed cyclization of diaryl- and alkyl aryl-1,3-dienes. In this approach, treatment of symmetric or unsymmetric diaryl- and alkyl aryl-1,3-dienes with a catalytic amount of trifluoromethanesulfonic acid gives a variety of indene derivatives in good to excellent yields under mild conditions.
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis