Merck
  • Rhizodegradation of petroleum hydrocarbons by Sesbania cannabina in bioaugmented soil with free and immobilized consortium.
Rhizodegradation of petroleum hydrocarbons by Sesbania cannabina in bioaugmented soil with free and immobilized consortium.
Journal of hazardous materials (2012-09-15)
Farhana Maqbool, Zhenyu Wang, Ying Xu, Jian Zhao, Dongmei Gao, Yang-Guo Zhao, Zulfiqar A Bhatti, Baoshan Xing
ABSTRACT
The present study reports the effect of bioaugmentation by free and immobilized bacterial culture on the rhizodegradation of petroleum-polluted soil using Sesbania cannabina plant. Total petroleum hydrocarbon (TPH), hydrocarbon-degrading bacterial counts, microbial activity and root morphology were assessed during 120 days of plant growth. TPH concentration analyzed by GC-MS showed that bioaugmentation did not improve the TPH degradation. TPH concentration decreased from 2541 mg kg(-1) to 673 mg kg(-1) and 867 mg kg(-1) in the rhizosphere of free (FR) and immobilized bacterial inoculated (IR) soil, respectively at the 120th day while in the rhizosphere of uninoculated soil (CR) concentration decreased to 679 mg kg(-1) only at the 90th day, showing higher and rapid rhizodegradation with indigenous bacteria than bioaugmented bacterial cultures. Various predominant bacterial groups responsible for higher TPH degradation in the rhizosphere of S. cannabina were identified by PCR-DGGE analysis. It is concluded that natural plant-microbe interaction in the rhizosphere of S. cannabina was efficient enough to degrade TPH and plant rhizosphere keeps bacterial community in its surrounding therefore immobilized culture had no obvious effect on petroleum degradation.
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined