Merck
Mercury speciation analysis in terrestrial animal tissues.
Talanta (2012-09-13)
J J Berzas Nevado, R C Rodríguez Martín-Doimeadios, F J Guzmán Bernardo, N Rodríguez Fariñas, M J Patiño Ropero
ABSTRACT
No previous analytical procedures are available and validated for mercury speciation analysis in terrestrial animal tissues. This analysis is a difficult task both because the expected concentrations are low, since important accumulation process are not likely to occur, and also because there are not commercially available certified reference material. Thus, an analytical methodology has been developed and validated for mercury speciation for the specific case of terrestrial animal tissues. The proposed method is based on the quantitative extraction of the species by closed-vessel microwave assisted heating with an alkaline reagent, followed by ethylation. The ethylated derivatives were then submitted to head-space solid phase microextraction with a 100 μm polidimethylsiloxane-coated fiber, and desorbed onto a gas chromatograph coupled to atomic fluorescence detection via pyrolysis unit (HS-SPME-GC-pyro-AFS). Procedural detection limits were 31.8 ng g(-1) and 52.5 ng g(-1) for CH(3)Hg(+) and Hg(2+), respectively, for liver and 35.3 ng g(-1) and 58.1 ng g(-1) for CH(3)Hg(+) and Hg(2+), respectively, for kidney. These limits of detection are 5.5 and 6 times better than the obtained without solid phase microextraction for CH(3)Hg(+) and Hg(2+), respectively. The methodology was found linear up to 120 μg L(-1) and reproducible from one day to the following. It was validated with certified reference materials NCS ZC 71001 (beef liver) and BCR No 186 (pig kidney) for total mercury, calculated as the sum of species, and with spiked red deer liver and kidney for speciation. Finally, it was applied to the analysis of samples of red deer liver, red deer kidney and wild boar kidney coming from the Almadén's mercury mining area (Ciudad Real, Spain), the longest and largest producer of mercury in the world until its closure in 2002.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium bromide, 98%