Merck
  • Involvement of presynaptic voltage-dependent Kv3 channel in endothelin-1-induced inhibition of noradrenaline release from rat gastric sympathetic nerves.
Involvement of presynaptic voltage-dependent Kv3 channel in endothelin-1-induced inhibition of noradrenaline release from rat gastric sympathetic nerves.
European journal of pharmacology (2012-09-12)
Kumiko Nakamura, Takahiro Shimizu, Kenjiro Tanaka, Keisuke Taniuchi, Kunihiko Yokotani
ABSTRACT
We previously reported that two types of K(+) channels, the BK type Ca(2+)-activated K(+) channel coupled with phospholipase C (PLC) and the voltage-dependent K(+) channel (Kv channel), are, respectively, involved in the prostanoid TP receptor- and muscarinic M(2) receptor-mediated inhibition of noradrenaline (NA) release from rat gastric sympathetic nerves. In the present study, therefore, we examined whether these K(+) channels are involved in endothelin-1-induced inhibition of NA release, using an isolated, vascularly perfused rat stomach. The gastric sympathetic postganglionic nerves around the left gastric artery were electrically stimulated twice at 2.5 Hz for 1 min, and endothelin-1 was added during the second stimulation. Endothelin-1 (1, 2 and 10 nM) dose-dependently inhibited gastric NA release. Endothelin-1 (2 nM)-induced inhibition of NA release was neither attenuated by PLC inhibitors [U-73122 (3 μM) and ET-18-OCH(3) (3 μM)] nor by Ca(2+)-activated K(+) channel blockers [charybdotoxin (0.1 μM) (a blocker of BK type K(+) channel) and apamin (0.3 μM) (a blocker of SK type K(+) channel)]. The endothelin-1-induced inhibitory response was also not attenuated by α-dendrotoxin (0.1 μM) (a selective inhibitor of Kv1 channel), but abolished by 4-aminopyridine (20 μM) (a selectively inhibitory dose for Kv3 channel). These results suggest the involvement of a voltage-dependent Kv3 channel in the endothelin-1-induced inhibition of NA release from the gastric sympathetic nerves in rats.
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)
Sigma-Aldrich
Charybdotoxin, ≥90% (HPLC)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)