Merck
  • Silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with pyridyne.
Silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with pyridyne.
Organic & biomolecular chemistry (2012-09-06)
Lingyong Jiang, Xingxin Yu, Bing Fang, Jie Wu
ABSTRACT
A silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with pyridyne is presented. Different outcomes are obtained, depending on the pyridynes utilized in the transformation.
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis