Merck
  • Revision of iron(III)-citrate speciation in aqueous solution. Voltammetric and spectrophotometric studies.
Revision of iron(III)-citrate speciation in aqueous solution. Voltammetric and spectrophotometric studies.
Analytica chimica acta (2012-09-04)
Petra Vukosav, Marina Mlakar, Vladislav Tomišić
ABSTRACT
A detailed study of iron (III)-citrate speciation in aqueous solution (θ=25°C, I(c)=0.7 mol L(-1)) was carried out by voltammetric and UV-vis spectrophotometric measurements and the obtained data were used for reconciled characterization of iron (III)-citrate complexes. Four different redox processes were registered in the voltammograms: at 0.1 V (pH=5.5) which corresponded to the reduction of iron(III)-monocitrate species (Fe:cit=1:1), at about -0.1 V (pH=5.5) that was related to the reduction of FeL(2)(5-), FeL(2)H(4-) and FeL(2)H(2)(3-) complexes, at -0.28 V (pH=5.5) which corresponded to the reduction of polynuclear iron(III)-citrate complex(es), and at -0.4V (pH=7.5) which was probably a consequence of Fe(cit)(2)(OH)(x) species reduction. Reversible redox process at -0.1 V allowed for the determination of iron(III)-citrate species and their stability constants by analyzing E(p) vs. pH and E(p) vs. [L(4-)] dependence. The UV-vis spectra recorded at varied pH revealed four different spectrally active species: FeLH (logβ=25.69), FeL(2)H(2)(3-) (log β=48.06), FeL(2)H(4-) (log β=44.60), and FeL(2)(5-) (log β=38.85). The stability constants obtained by spectrophotometry were in agreement with those determined electrochemically. The UV-vis spectra recorded at various citrate concentrations (pH=2.0) supported the results of spectrophotometric-potentiometric titration.
Product Description
Supelco
Iron(III) citrate tribasic monohydrate, 18-20% Fe basis (T)
Sigma-Aldrich
Iron(III) citrate, technical grade
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture