Merck
  • Fluorescence enhancement of CdTe/CdS quantum dots by coupling of glyphosate and its application for sensitive detection of copper ion.
Fluorescence enhancement of CdTe/CdS quantum dots by coupling of glyphosate and its application for sensitive detection of copper ion.
Analytica chimica acta (2012-09-04)
Zhengqing Liu, Shaopu Liu, Pengfei Yin, Youqiu He
ABSTRACT
A novel fluorescent probe for Cu(2+) determination based on the fluorescence quenching of glyphosate (Glyp)-functionalized quantum dots (QDs) was firstly reported. Glyp had been used to modify the surface of QDs to form Glyp-functionalized QDs following the capping of thioglycolic acid on the core-shell CdTe/CdS QDs. Under the optimal conditions, the response was linearly proportional to the concentration of Cu(2+) between 2.4×10(-2)μg mL(-1) and 28μg mL(-1), with a detection limit of 1.3×10(-3)μg mL(-1) (3δ). The Glyp-functionalized QDs fluorescent probe offers good sensitivity and selectivity for detecting Cu(2+). The fluorescent probe was successfully used for the determination of Cu(2+) in environmental samples. The mechanism of reaction was also discussed.
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Supelco
Thioglycolic acid solution, ~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Sigma-Aldrich
Thioglycolic acid, ≥97%
Sigma-Aldrich
Thioglycolic acid, ≥99%
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
Sodium thioglycolate, ≥96.5% (iodometric)