Merck
  • A chemiluminescence array sensor based on graphene-magnetite-molecularly imprinted polymers for determination of benzenediol isomers.
A chemiluminescence array sensor based on graphene-magnetite-molecularly imprinted polymers for determination of benzenediol isomers.
Analytica chimica acta (2012-09-01)
Huamin Qiu, Chuannan Luo, Min Sun, Fuguang Lu, Lulu Fan, Xiangjun Li
ABSTRACT
A chemiluminescence (CL) array sensor for determination of benzenediol isomers simultaneously using the system of luminol-NaOH-H(2)O(2) based on a graphene-magnetite-molecularly imprinted polymer (GM-MIP) is described. Use of graphene in the GM-MIP thus prepared is helpful to improve the adsorption capacity, while use of magnetite nanoparticles can facilitate the isolation of GM-MIP at end of their synthesis, and rendering easier the use of the polymers in the array sensor. The adsorption performance and properties were characterized. The GM-MIP was used to increase the selectivity in CL analysis. In addition, the sensor was reusable and of good selectivity and adsorption capacity. The array sensor was finally used for the determination of hydroquinone, resorcinol and catechol in waste water samples simultaneously.
Product Description
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, ≥98%, FG