Merck
  • Co-delivery of paclitaxel and survivin shRNA by pluronic P85-PEI/TPGS complex nanoparticles to overcome drug resistance in lung cancer.
Co-delivery of paclitaxel and survivin shRNA by pluronic P85-PEI/TPGS complex nanoparticles to overcome drug resistance in lung cancer.
Biomaterials (2012-08-23)
Jianan Shen, Qi Yin, Lingli Chen, Zhiwen Zhang, Yaping Li
ABSTRACT
Drug resistance is a main obstacle for the successful chemotherapy of lung cancer. In this work, a new co-delivery system, P85-PEI/TPGS/PTX/shSur complex nanoparticles (PTPNs), to overcome paclitaxel (PTX) resistance in A549 human lung cancer was designed and developed. The experimental results showed that PTPNs could facilitate drug into cells and induce survivin shRNA (shSur) into nuclei on A549 and A549/T cells, achieve efficient gene delivery and induce availably RNA interference on A549/T cells. The IC(50) of PTPNs against A549/T cells was 360-fold lower than that of free PTX. The enhanced efficacy of PTPNs against A549/T cells was associated with PTX-induced apoptosis and cell arrest in G2/M phase. Down-regulation of survivin protein by PTPNs could lower the apoptosis threshold of drug resistant cells and render chemotherapeutic agents more effective. Moreover, the inhibition of GST activity by P85 was found to increase PTX accumulation in A549/T cells. The in vivo antitumor efficacy showed that PTPNs were more effective than that of the Taxol. As a result, the co-delivery of PTX and shSur by PTPNs could be a very powerful approach to improve the therapeutic effect of PTX in resistant lung cancer.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
D-α-Tocopherol polyethylene glycol 1000 succinate, BioXtra, water soluble vitamin E conjugate
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol)
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Poloxamer 188, solid