Merck
  • A peptide receptor-based bioelectronic nose for the real-time determination of seafood quality.
A peptide receptor-based bioelectronic nose for the real-time determination of seafood quality.
Biosensors & bioelectronics (2012-08-21)
Jong Hyun Lim, Juhun Park, Jung Ho Ahn, Hye Jun Jin, Seunghun Hong, Tai Hyun Park
ABSTRACT
We herein report a peptide receptor-based bioelectronic nose (PRBN) that can determine the quality of seafood in real-time through measuring the amount of trimethylamine (TMA) generated from spoiled seafood. The PRBN was developed using single walled-carbon nanotube field-effect transistors (SWNT-FETs) functionalized with olfactory receptor-derived peptides (ORPs) which can recognize TMA and it allowed us to sensitively and selectively detect TMA in real-time at concentrations as low as 10fM. Utilizing these properties, we were able to not only determine the quality of three kinds of seafood (oyster, shrimp, and lobster), but were also able to distinguish spoiled seafood from other types of spoiled foods without any pretreatment processes. Especially, the use of small synthetic peptide rather than the whole protein allowed PRBNs to be simply manufactured through a single-step process and to be reused with high reproducibility due to no requirement of lipid bilayers. Furthermore, the PRBN was produced on a portable scale making it effectively useful for the food industry where the on-site measurement of seafood quality is required.
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylamine hydrochloride, 98%