Merck
  • Folate-PEG-appended dendrimer conjugate with α-cyclodextrin as a novel cancer cell-selective siRNA delivery carrier.
Folate-PEG-appended dendrimer conjugate with α-cyclodextrin as a novel cancer cell-selective siRNA delivery carrier.
Molecular pharmaceutics (2012-08-10)
Hidetoshi Arima, Ayumi Yoshimatsu, Haruna Ikeda, Ayumu Ohyama, Keiichi Motoyama, Taishi Higashi, Akira Tsuchiya, Takuro Niidome, Yoshiki Katayama, Kenjiro Hattori, Tomoko Takeuchi
ABSTRACT
We previously reported that of the various polyamidoamine (PAMAM) STARBURST dendrimer (generation 3, G3) (dendrimer) conjugates with cyclodextrins (CyDs), the dendrimer (G3) conjugate with α-CyD having an average degree of substitution of 2.4 (α-CDE (G3)) has the greatest potential for a novel carrier for siRNA in vitro and in vivo. To improve the siRNA transfer activity and the lack of target specificity of α-CDE (G3), we prepared folate-polyethylene glycol (PEG)-appended α-CDEs (G3) (Fol-PαCs) with various degrees of substitution of folate (DSF) and evaluated their siRNA transfer activity to folate receptor (FR)-overexpressing cancer cells in vitro and in vivo. Of the three Fol-PαCs (G3, DSF 2, 4 and 7), Fol-PαC (G3, DSF 4) had the highest siRNA transfer activity in KB cells (FR-positive). Fol-PαC (G3, DSF 4) was endocytosed into KB cells through FR. No cytotoxicity of the siRNA complex with Fol-PαC (G3, DSF 4) was observed in KB cells (FR-positive) or A549 cells (FR-negative) up to the charge ratio of 100/1 (carrier/siRNA). In addition, the siRNA complex with Fol-PαC (G3, DSF 4) showed neither interferon response nor inflammatory response. Importantly, the siRNA complex with Fol-PαC (G3, DSF 4) tended to show the in vivo RNAi effects after intratumoral injection and intravenous injection in tumor cells-bearing mice. The FITC-labeled siRNA and TRITC-labeled Fol-PαC (G3, DSF 4) were actually accumulated in tumor tissues after intravenous injection in the mice. In conclusion, the present results suggest that Fol-PαC (G3, DSF 4) could potentially be used as a FR-overexpressing cancer cell-selective siRNA delivery carrier in vitro and in vivo.
Sigma-Aldrich
α-Cyclodextrin, ≥98%
Sigma-Aldrich
α-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
α-Cyclodextrin, purum, ≥98.0% (HPLC)
Sigma-Aldrich
α-Cyclodextrin, Wacker Chemie AG, 98.0-101.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
α-Cyclodextrin, Wacker Chemie AG, ≥98.0% (HPLC)