Merck
  • Inter-tissue networks between the basal forebrain, hippocampus, and prefrontal cortex in a model for depression caused by disturbed sleep.
Inter-tissue networks between the basal forebrain, hippocampus, and prefrontal cortex in a model for depression caused by disturbed sleep.
Journal of neurogenetics (2012-07-13)
Markus Lagus, Natalia Gass, Juha Saharinen, Sergey Savelyev, Tarja Porkka-Heiskanen, Tiina Paunio
ABSTRACT
Disturbances in sleep are encountered in the majority of patients with depressive disorder. To elucidate the molecular mechanisms behind this relationship, we examined gene expression changes in a rodent model for disturbed sleep and depression. The animals were treated with daily injections of clomipramine to affect their sleep during early infancy. This early interference with sleep is known to induce depression-like behavior in adult animals. After 2 weeks of treatment, the change in gene expression was examined using the Affymetrix Rat 230.2 chip. We studied the gene expression in the basal forebrain, hippocampus, and frontal cortex and combined the results to reveal the otherwise indissectible networks between and around the tissues. The major disrupted pathways between the three brain areas were related to synaptic transmission, regulation of translation, and ubiquitinylation. The involved pathways were within the cellular components of the axons, growth cones, melanosomes, and pigment granules. A network analysis allowing for additional interactors, in the form of chemicals or gene products, revealed a disturbed communicational network between the different brain areas. This disturbed network is centered around serotonin, Mn(II), and Rhoa. The findings elucidate inter-tissue pathways and networks in the brain that are involved in sleep and mood regulation. The findings are of uttermost interest, some are quite predictable and obvious, but some are novel or have only been proposed by rare theoretical speculations (such as the melanosome and Mn(II) involvement). Equally important as the findings are the methods described in this article. In this study, we present two novel simple ways to perform system biological analysis based on gene expression array data. We used two already existing tools in a new way, and by careful planning of the input data, managed to extrapolate intricate hidden inter-tissue networks to build a molecular picture of disease.
Product Description
Supelco
Clomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Clomipramine hydrochloride, ≥98% (HPLC), powder