Merck
  • Fabrication of magnetic Ni nanoparticles functionalized water-soluble graphene sheets nanocomposites as sorbent for aromatic compounds removal.
Fabrication of magnetic Ni nanoparticles functionalized water-soluble graphene sheets nanocomposites as sorbent for aromatic compounds removal.
Journal of hazardous materials (2012-06-29)
Shuwen Li, Zhiyong Niu, Xing Zhong, Honglei Yang, Yaojie Lei, Fengwei Zhang, Wuquan Hu, Zhengping Dong, Jun Jin, Jiantai Ma
ABSTRACT
The magnetic Ni nanoparticles functionalized water-soluble graphene sheets nanocomposites (Ni@GSs-C(CH(3))(2)COONa) were fabricated via a facile and mild strategy. We designed a simple and efficient approach (the addition of cyano radicals) to improve the water solubility of nanocomposites. The Ni@GSs-C(CH(3))(2)COONa nanocomposites had great potential as an effective absorbent for removing aromatic compounds from waste water owing to their rapid absorption rate, high absorption capacity, convenient magnetic separation and re-use property.
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene