Merck
  • Rapid separations of nile blue stained microorganisms as cationic charged species by chip-CE with LIF.
Rapid separations of nile blue stained microorganisms as cationic charged species by chip-CE with LIF.
Electrophoresis (2012-06-01)
Nantana Nuchtavorn, Fritz Bek, Mirek Macka, Worapot Suntornsuk, Leena Suntornsuk
ABSTRACT
Rapid detection of microorganisms by alternative methods is desirable. Electromigration separation methods have the capability to separate microorganisms according to their charge and size and laser-induced fluorescence (LIF) detection have single-cell detection capability. In this work, a new combined separation and detection scheme was introduced using chip-based capillary electrophoresis (chip-CE) platform with LIF detection. Three microorganisms Escherichia coli, Staphylococcus aureus, and Candida albicans were selected as representatives of Gram-positive bacteria, Gram-negative bacteria, and fungi. While their cells carry an overall negative charge in neutral to alkaline pH, staining them with nile blue (NB) provided highly sensitive LIF detection with excitation and emission wavelengths at 635 nm and 685 nm, respectively, and at the same time, the overall charge was converted to positive. Electrolyte pH and concentration of polyethylene oxide (PEO) significantly affected the resolution of the microorganisms. Their optimal separation in the 14 mm separation channel was achieved in less than 30 s (R(s) > 5.3) in an electrolyte consisting of 3.94 mM Tris, 0.56 mM boric acid, 0.013 mM ethylenediaminetetraacetic acid disodium salt dihydrate (pH 10.5), and 0.025% PEO, with injection/separation voltages of +1000/+1000 V. The separation mechanism is likely employing contributions to the overall cationic charge from both the prevalently anionic membrane proteins and the cationic NB. Importantly, the resulting cationic NB-stained cells exhibited excellent separation selectivity and efficiency of ∼38000 theoretical plates for rapid separations within 30-40 s. The results indicate the potential of chip-CE for microbial analysis, which offers separations of a wide range of species with high efficiency, sensitivity, and throughput.
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %