Merck
  • Kupffer cells suppress perfluorononanoic acid-induced hepatic peroxisome proliferator-activated receptor α expression by releasing cytokines.
Kupffer cells suppress perfluorononanoic acid-induced hepatic peroxisome proliferator-activated receptor α expression by releasing cytokines.
Archives of toxicology (2012-06-01)
Xuemei Fang, Shanshan Zou, Yuanyuan Zhao, Ruina Cui, Wei Zhang, Jiayue Hu, Jiayin Dai
ABSTRACT
Kupffer cells (KCs) have been demonstrated to play a role in the regulation of intra-hepatic lipid metabolism through the synthesis and secretion of biologically active products. The involvement of KCs in the disturbance of lipid metabolism that induced by perfluorononanoic acid (PFNA), a known agonist of the peroxisome proliferator-activated receptor alpha (PPARα), was investigated in this study. Rats were exposed to PFNA or PFNA combined with gadolinium chloride, an inhibitor of KCs, for 14 days. PFNA exposure dose-dependently increased absolute and relative liver weights, induced triglyceride accumulation, up-regulated the expression of both SERBP-1c and PPARα, and stimulated the release of TNFα and IL-1β. Inactivation of KCs markedly lowered TNFα and IL-1β level, enhanced PFNA-induced expression of PPARα and its target genes, and reduced liver triglyceride levels. In vitro, PFNA-induced expression of PPARα in primary cultured hepatocytes was suppressed by recombinant rat TNFα and IL-1β. However, inhibition of the NF-κB pathway prevented this. Transient transfection and promoter analysis further revealed that these two cytokines and NF-κB were coordinately involved in the suppression of PPARα promoter activity. Our data demonstrate that TNFα and IL-1β released from KCs following PFNA exposure can suppress the expression of PPARα via NF-κB pathway, which partially contribute to the evident accumulation of triglycerides in rat liver.
Sigma-Aldrich
Perfluorononanoic acid, 97%
Sigma-Aldrich
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis