Merck
  • Preparation and evaluation of folate-modified cationic pluronic micelles for poorly soluble anticancer drug.
Preparation and evaluation of folate-modified cationic pluronic micelles for poorly soluble anticancer drug.
Drug delivery (2012-05-31)
Wei Xu, Yanan Cui, Peixue Ling, Ling-bing Li
ABSTRACT
The aim of this study was to construct novel targeting polymeric micelles. Folate-Poly (ethylenimine)-Pluronic copolymers were synthesized. A paclitaxel (PTX)-loaded mixed micelles consisting of Folate-Poly (ethylenimine)-Pluronic and Pluronic L121 copolymers have been developed. The mixed micelles showed nano-sized spherical morphology. The solubilization capacity of the mixed micelles was higher than Folate-Poly (ethylenimine)-Pluronic micelles because L121 has high solubilization capacity. MTT colorimetric test revealed that PTX in Folate-Poly (ethylenimine)-Pluronic micelles demonstrated the maximum anticancer activity. Pluronic-poly (ethylenimine) micelles and folate-modified Pluronic-poly(ethylenimine) micelles showed a marked increase of cellular accumulation compared with Pluronic P123 micelles. The biodistribution and retention of intravenously (i.v.) administered micelles to rats were determined. Folate-Poly (ethylenimine)-Pluronic micelles demonstrated enhanced pulmonary retention in rats after injection when compared to Pluronic P123 micelles.
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800