- Photoswitched protein adsorption on electrostatically self-assembled azobenzene films.
Photoswitched protein adsorption on electrostatically self-assembled azobenzene films.
Chemphyschem : a European journal of chemical physics and physical chemistry (2012-05-24)
Jian Zhang, Te Hu, Yibiao Liu, Yanxuan Ma, Jie Dong, Liping Xu, Yudong Zheng, Huai Yang, Guojie Wang
PMID22618888
ABSTRACT
Photoresponsive polymeric films fabricated by a facile electrostatic self-assembly technique are utilized to switch protein adsorption by light irradiation. The introduction of SiO(2) nanoparticles on the substrate results in a large reversible change of both wettability and protein adsorption.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O