Merck
  • Enantiomeric derivatives of tokinolide B: absolute configuration and biological properties.
Enantiomeric derivatives of tokinolide B: absolute configuration and biological properties.
Journal of natural products (2012-05-12)
Alejandra León, J Antonio Cogordán, Olov Sterner, Guillermo Delgado
ABSTRACT
The enantiomeric lactams (-)-8, (+)-8, (+)-9, and (-)-9 were formed by the reaction of the dimeric phthalide rac-tokinolide B (rac-3) with (R)-(+)-α-methylbenzylamine and (S)-(-)-α-methylbenzylamine. The absolute configurations of compounds 8 and 9 were assigned by experimental and theoretically calculated electronic circular dichroism methods for (+)-8 and (-)-9. Compounds 3, 5, (-)-8, (+)-8, (+)-9, and (-)-9 displayed cytotoxic activity toward several human tumor cell lines, with (-)-8 and (-)-9 being the most potent.
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, purum, ≥98.0% (sum of enantiomers, GC)
Supelco
(R)-(+)-α-Methylbenzylamine, derivatization grade (chiral), LiChropur, ≥99.0%
Sigma-Aldrich
α-Methylbenzylamine, 99%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, ChiPros®, produced by BASF, ≥99.0%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamine, 98%