Merck
  • Gas phase chemistry of Li+ with amides: the observation of LiOH loss in mass spectrometry.
Gas phase chemistry of Li+ with amides: the observation of LiOH loss in mass spectrometry.
Journal of the American Society for Mass Spectrometry (2012-05-03)
Cheng Guo, Yuping Zhou, Pengyuan Liu, Yunfeng Chai, Yuanjiang Pan
ABSTRACT
Collision-induced dissociation (CID) of Li(+) adducts of three sets of compounds that contains an amide bond, including 2-(4, 6-dimethoxypyrimidin-2-ylsulfanyl)-N-phenylbenzamide, its derivatives and simpler structures was investigated by electrospray ionization tandem mass spectrometry (ESI-MS/MS). Observed fragment ions include those that reflect loss of LiOH. Other product ions result from the Smiles rearrangement and direct C-S bond cleavage. MS/MS of H/D exchange products demonstrated occurrence of a 1,3-H shift from the amide nitrogen atom to the phenyl ring of these compounds. The LiOH loss from Li(+) adducts of amides was further examined by CID of [M + Li](+) ions of N-phenylbenzamide and N-phenylcinnamide. Loss of LiOH was essentially the sole fragmentation reaction observed for the former. For the latter, both losses of LiOH and H(2)O were discovered. The presence of electron-donating substituents of the phenyl ring of these compounds was found to facilitate elimination of LiOH, while that loss was retarded by electron-withdrawing substituents. Proposed fragment ion structures were supported by elemental compositions deduced from ultrahigh resolution Fourier transform ion cyclotron resonance tandem mass spectrometry (FTICR-MS/MS) m/z value determinations. Density functional theory-based (DFT) calculations were performed to evaluate potential mechanisms for these reactions.
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide ChemBeads
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%