Merck
  • Diagnosing vaginal infections through measurement of biogenic amines by ion mobility spectrometry.
Diagnosing vaginal infections through measurement of biogenic amines by ion mobility spectrometry.
European journal of obstetrics, gynecology, and reproductive biology (2012-04-24)
Jack D Sobel, Zeev Karpas, Avraham Lorber
ABSTRACT
To compare diagnosis of bacterial vaginosis according to the Amsel criteria with measurement by ion mobility spectrometry (IMS) of the biogenic amines that are present in vaginal discharge fluid. Duplicate samples of vaginal fluid were collected from 115 unselected and consecutive patients in a vaginitis clinic in Detroit. All samples were evaluated using Amsel criteria and the results were compared with the diagnosis based on the IMS results. The incidence rate of vaginal infections was assessed on the basis of both tests and the frequency of BV was found to be 17.4%. The sensitivity and specificity for bacterial vaginosis diagnosis using IMS determination were 95.5% and 98.9%, respectively, with an accuracy of 94.4%. The results show that IMS may be used to rapidly diagnose this common vaginal infection with high accuracy.
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Trimethylamine hydrochloride, 98%