Merck
  • Plasma-Lyte 148 vs 0.9% saline for fluid resuscitation in diabetic ketoacidosis.
Plasma-Lyte 148 vs 0.9% saline for fluid resuscitation in diabetic ketoacidosis.
Journal of critical care (2012-03-24)
Horng-Ruey Chua, Balasubramanian Venkatesh, Edward Stachowski, Antoine G Schneider, Kelly Perkins, Suzy Ladanyi, Peter Kruger, Rinaldo Bellomo
ABSTRACT
The purpose of the study was to determine the effects of Plasma-Lyte 148 (PL) vs 0.9% saline (NS) fluid resuscitation in diabetic ketoacidosis (DKA). A multicenter retrospective analysis of adults admitted for DKA to the intensive care unit, who received almost exclusively PL or NS infusion up until 12 hours, was performed. Nine patients with PL and 14 patients with NS were studied. Median serum bicarbonate correction was higher in the PL vs NS groups at 4 to 6 hours (8.4 vs 1.7 mEq/L) and 6 to 12 hours (12.8 vs 6.2 mEq/L) from baseline (P < .05). Median standard base excess improved by 10.5 vs 4.2 mEq/L at 4 to 6 hours and by 16.0 vs 9.1 mEq/L at 6 to 12 hours in the PL and NS groups, respectively (P < .05). Chloride levels increased significantly in the NS vs PL groups over 24 hours. Potassium levels were lower at 6 to 12 hours in the PL group. Mean arterial blood pressure was higher at 2 to 4 hours in the PL group, whereas cumulative urine output was lower at 4 to 6 hours in the NS group. There were no differences in glycemic control or duration of intensive care unit stay. Patients with DKA resuscitated with PL instead of NS had faster initial resolution of metabolic acidosis and less hyperchloremia, with a transiently improved blood pressure profile and urine output.
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%