Merck
  • A POSS-based supramolecular amphiphile and its hierarchical self-assembly behaviors.
A POSS-based supramolecular amphiphile and its hierarchical self-assembly behaviors.
Macromolecular rapid communications (2012-03-15)
Binbin Jiang, Wei Tao, Xin Lu, Yong Liu, Haibao Jin, Yan Pang, Xiaoyi Sun, Deyue Yan, Yongfeng Zhou
ABSTRACT
A polyhedral oligomeric silsesquioxane (POSS)-based supramolecular amphiphile is prepared from the host-guest inclusion complexation between a mono adamantane-functionalized POSS (AD-POSS) and a β-cyclodextrin oligomer (P(β-CD)). Assisted by the interface of H(2)O/toluene, the obtained supramolecular hybrids self-assemble into stable hollow nanospheres with thick walls. These hollow nanospheres aggregate together into a sphere layer through a spin coating technique, which then further transforms into a thin porous film containing nanometer-scale holes. The hollow nanospheres have a low cytotoxicity. The in vitro cell culture indicates the nanoporous films promote adhesion and proliferation of cells. The self-assembly morphologies and structures have been carefully characterized by SEM, TEM, AFM, DLS, XPS and water-contact angle measurements, and the self-assembly mechanism has also been discussed.
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%
Sigma-Aldrich
(±)-Epichlorohydrin, purum, ≥99% (GC)
Supelco
(±)-Epichlorohydrin, analytical standard