Merck
Induced circular dichroism of stereoregular vinyl polymers.
Chemical communications (Cambridge, England) (2012-03-08)
Lung-Chi Chen, Yung-Cheng Mao, Shih-Chieh Lin, Ming-Chia Li, Rong-Ming Ho, Jing-Cherng Tsai
ABSTRACT
Stereoregular vinyl polymers, poly(2-vinyl pyridine)s (P2VPs), were synthesized to examine the tacticity effect on the induced circular dichroism (ICD) via association with chiral acids. The ICD was found to be strongly dependent on the isotacticity of the P2VPs and the acidity of chiral acid in addition to its bulkiness.
Product Description
Supelco
Poly(2-vinylpyridine), analytical standard, average Mw 159,000 (Typical), average Mn 152,000 (Typical)
Supelco
Poly(2-vinylpyridine), analytical standard, average Mw 37,500 (Typical), average Mn 35,000 (Typical)