Merck
  • Role of dissolved oxygen availability on lactobionic acid production from whey by Pseudomonas taetrolens.
Role of dissolved oxygen availability on lactobionic acid production from whey by Pseudomonas taetrolens.
Bioresource technology (2012-02-09)
Saúl Alonso, Manuel Rendueles, Mario Díaz
ABSTRACT
The influence of dissolved oxygen availability on cell growth and lactobionic acid production from whey by Pseudomonas taetrolens has been investigated for the first time. Results from pH-shift bioreactor cultivations have shown that high agitation rate schemes stimulated cell growth, increased pH-shift values and the oxygen uptake rate by cells, whereas lactobionic acid production was negatively affected. Conversely, higher aeration rates than 1.5 Lpm neither stimulated cell growth nor lactobionic acid production (22% lower for an aeration rate of 2 Lpm). Overall insights into bioprocess performance enabled the implementation of 350 rpm as the optimal agitation strategy during cultivation, which increased lactobionic productivity 1.2-fold (0.58-0.7 g/Lh) compared to that achieved at 1000 rpm. Oxygen supply has been shown to be a key bioprocess parameter for enhanced overall efficiency of the system, representing essential information for the implementation of lactobionic acid production at a large scale.
Product Description
Lactobionic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lactobionic acid, 97% (TLC)
Sigma-Aldrich
Lactobionic acid, ≥97% (TLC), cell impermeant agent